Learn More
2,2',5,5'-Tetramethylbiphenyl, 98%
Product Code. p-7046159
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11348046 1 g 1g
11358046 5 g 5g
2 options
Product Code. 11348046
Brand: Thermo Scientific Alfa Aesar L13330.03
Request Bulk or Custom Format
CAS: 3075-84-1 | C16H18 | 210.32 g/mol
2,2',5,5'-Tetramethylbiphenyl is used to make other chemicals and as an intermediate in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2′,5,5′-Tetramethylbiphenyl is used to make other chemicals and as an intermediate in pharmaceuticals.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3075-84-1
Molecular Formula C16H18
Molecular Weight (g/mol) 210.32
MDL Number MFCD00151846
InChI Key ZHTROMYSDSTCCE-UHFFFAOYSA-N
PubChem CID 137818
IUPAC Name 2-(2,5-dimethylphenyl)-1,4-dimethylbenzene
SMILES CC1=CC=C(C)C(=C1)C1=CC(C)=CC=C1C
Specifications
Melting Point 48°C to 51°C
Boiling Point 284°C (732 mmHg)
Quantity 1 g
Beilstein 2046200
Synonym 2,2',5,5'-tetramethylbiphenyl, 2,2',5,5'-tetramethyl-1,1'-biphenyl, 1,1'-biphenyl, 2,2',5,5'-tetramethyl, biphenyl-, 2,2',5,5'-tetramethyl, 2-2,5-dimethylphenyl-1,4-dimethylbenzene, acmc-20aokf, 2,5,2',5'-tetramethylbiphenyl, 1,1'-biphenyl,2,2',5,5'-tetramethyl, 2,2',5,5'-tetramethyl-1,1'-biphenyl #
Solubility Information Insoluble in water.
Formula Weight 210.32
Percent Purity 98%
Chemical Name or Material 2,2',5,5'-Tetramethylbiphenyl