Learn More
2-(2,4,5-Trichlorophenoxy)propionic acid, 96%, Thermo Scientific™
Product Code. 11397995
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397995 5 g 5g
11308005 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11397995 Supplier Thermo Scientific Alfa Aesar Supplier No. L08136.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 93-72-1
Molecular Formula C9H7Cl3O3
Molecular Weight (g/mol) 269.50
MDL Number MFCD00002646
InChI Key ZLSWBLPERHFHIS-UHFFFAOYNA-N
Synonym fenoprop, silvex, 2-2,4,5-trichlorophenoxy propionic acid, double strength, fenormone, propon, color-set, aqua-vex, sta-fast, fruitone t
PubChem CID 7158
ChEBI CHEBI:34758
IUPAC Name 2-(2,4,5-trichlorophenoxy)propanoic acid
SMILES CC(OC1=CC(Cl)=C(Cl)C=C1Cl)C(O)=O
Specifications
Melting Point 176°C to 180°C
Quantity 5 g
UN Number UN3077
Beilstein 1985768
Formula Weight 269.51
Percent Purity 96%
Chemical Name or Material 2-(2,4,5-Trichlorophenoxy)propionic acid