Learn More
2,2,4,4-Tetramethyl-1,3-cyclobutanediol, TRC
Product Code. 30451854
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451854 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30451854 Supplier Toronto Research Chemicals Supplier No. TRCT889765500MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,4-Dihydroxy-1,1,3,3-tetramethylcyclobutane
CAS 3010-96-6
Molecular Formula C8 H16 O2
Quantity 500 mg
Synonym 2,2,4,4-Tetramethyl-1,3-cyclobutanediol, 1,3-Dihydroxy-2,2,4,4-tetramethylcyclobutane, 2,2,4,4-Tetramethyl-1,3-cyclobutandiol, 2,4-Dihydroxy-1,1,3,3-tetramethylcyclobutane, NSC 46473, Tetramethyl-1,3-cyclobutanediol, 1,3-Cyclobutanediol','2,2,4,4-tetramethyl-
InChI Formula InChI=1S/C8H16O2/c1-7(2)5(9)8(3,4)6(7)10/h5-6,9-10H,1-4H3
SMILES CC1(C)C(O)C(C)(C)C1O
IUPAC Name 2,2,4,4-tetramethylcyclobutane-1,3-diol
Molecular Weight (g/mol) 144.211
Formula Weight 144.115