Learn More
2,2,3,3,4,4,4-Heptafluorobutanoic anhydride, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 336-59-4 |
| Molecular Formula | C8F14O3 |
| MDL Number | MFCD00000432 |
| Quantity | 1 g |
| SMILES | O=C(OC(C(F)(F)C(F)(F)C(F)(F)F)=O)C(F)(F)C(F)(F)C(F)(F)F |
| Molecular Weight (g/mol) | 410.06 |
| Percent Purity | 98% |
| Chemical Name or Material | 2,2,3,3,4,4,4-Heptafluorobutanoic anhydride |