Learn More
2,2,3,3,3-Pentafluoropropionic Acid, Sigma-Aldrich™
Specifications
Specifications
| CAS | 422-64-0 |
| Density | 1.57 g/cm3 at 20°C |
| Boiling Point | 96°C to 97°C/984 hPa |
| MDL Number | MFCD00004170 |
| Quantity | 10 mL |
| Synonym | 2,2,3,3,3-Pentafluoropropionic acid, Perfluoropropionic acid, 2,2,3,3,3-Pentafluoropropionic acid |
| Molecular Weight (g/mol) | 164.03 |
| Grade | Research |