Learn More
2,2,2-Trichloroethanol, Sigma-Aldrich™
Specifications
Specifications
| CAS | 115-20-8 |
| Melting Point | 17°C to 18°C |
| Density | 1.56 g/cm3 at 20°C |
| Boiling Point | 151°C to 153°C/1013 hPa |
| MDL Number | MFCD00004677 |
| Quantity | 100 mL |
| Synonym | 2,2,2-Trichloroethanol, β,β,β-Trichloroethyl alcohol, 2,2,2-Trichloroethanol |
| Molecular Weight (g/mol) | 149.4 |
| Grade | Research |