Learn More
2-(1H-Imidazol-1-yl)aniline, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 26286-54-4 |
| Molecular Formula | C9H9N3 |
| MDL Number | MFCD03274531 |
| Quantity | 5 g |
| SMILES | NC1=C(C=CC=C1)N1C=CN=C1 |
| Molecular Weight (g/mol) | 159.19 |
| Percent Purity | 95% |
| Chemical Name or Material | 2-(1H-Imidazol-1-yl)aniline |