Learn More
2,1,3-Benzoxadiazole-5-carboxylic acid, 97%, Thermo Scientific™
Product Code. 10427422
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10427422 250 mg 250mg
10501682 1 g 1g
10021694 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10427422 Supplier Thermo Scientific Maybridge Supplier No. CC09701CB
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Chemical Identifiers
CAS 19155-88-5
Molecular Formula C7H3N2O3
Molecular Weight (g/mol) 163.11
MDL Number MFCD00276977
InChI Key WZUFYJFTOVGJJT-UHFFFAOYSA-M
Synonym benzo c 1,2,5 oxadiazole-5-carboxylic acid, benzofurazan-5-carboxylic acid, 5-benzofurazancarboxylic acid, benzo c 1,2,5-oxadiazole-5-carboxylic acid, pubchem3995, maybridge1_002587, 4-bromo-2,6-dimethylanisole,, 2,1,3-benzoxadiazole-5-carboxylicacid, benzo 1,2,5 oxadiazole-5-carboxylic acid, benzo 2,1,3 oxadiazole-5-carboxylic acid
PubChem CID 2735454
IUPAC Name 2,1,3-benzoxadiazole-5-carboxylic acid
SMILES [O-]C(=O)C1=CC2=NON=C2C=C1
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 164.12
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzoxadiazole-5-carboxylic acid