Learn More
2-[1-(2-Naphthyl)ethyl]benzoic Acid, TRC
Product Code. 30477438
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477438 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30477438 Supplier Toronto Research Chemicals Supplier No. TRCN3720502G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-[1-(2-Naphthyl)ethyl]benzoic Acid
CAS 904236-46-0
Molecular Formula C19 H16 O2
Quantity 2 g
InChI Formula InChI=1S/C19H16O2/c1-13(17-8-4-5-9-18(17)19(20)21)15-11-10-14-6-2-3-7-16(14)12-15/h2-13H,1H3,(H,20,21)
SMILES CC(c1ccc2ccccc2c1)c3ccccc3C(=O)O
IUPAC Name 2-(1-naphthalen-2-ylethyl)benzoic acid
Molecular Weight (g/mol) 276.33
Formula Weight 276.115