Learn More
(1S)-(+)-Camphor-10-sulfonic Acid, Sigma-Aldrich™
Specifications
Specifications
| CAS | 3144-16-9 |
| Melting Point | 195°C (Decomposes) |
| MDL Number | MFCD00064157 |
| Quantity | 100 g |
| Synonym | (1S)-(+)-Camphor-10-sulfonic acid, for resolution of racemates, (1S)-(+)-Camphor-10-sulfonic acid |
| Molecular Weight (g/mol) | 232.3 |
| Grade | Research |