Learn More
(1S,2R)-2-(2-Benzylamino-1-hydroxyethyl)-6-fluorochromane, Mikromol™
Description
(1S,2R)-2-(2-Benzylamino-1-hydroxyethyl)-6-fluorochromane
Specifications
Specifications
| Chemical Name or Material | (1S,2R)-2-(2-Benzylamino-1-hydroxyethyl)-6-fluorochromane |
| CAS | 129050-27-7 |
| Analyte or Component Names | (1S,2R)-2-(2-Benzylamino-1-hydroxyethyl)-6-fluorochromane |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18H20FNO2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2H-1-Benzopyran-2-methanol, 6-fluoro-3,4-dihydro-α-[[(phenylmethyl)amino]methyl]-, (αS,2R)-, 2H-1-Benzopyran-2-methanol, 6-fluoro-3,4-dihydro-α-[[(phenylmethyl)amino]methyl]-, [R-(R*,S*)]-, (αS,2R)-6-Fluoro-3,4-dihydro-α-[[(phenylmethyl)amino]methyl]-2H-1-benzopyran-2-methanol, (-)-(S)-N-Benzyl-2-[(R)-6-fluorochroman-2-yl]-2-hydroxyethanamine, (1S)-1-((2R)-6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-yl)-2-[(phenylmethyl)amino]ethanol, 2H-1-Benzopyran-2-methanol 6-fluoro-3,4-dihydro-α-[[(phenylmethyl)amino]methyl]-, (αS,2R)-, (1S,2R)-2-(2-Benzylamino-1-hydroxyethyl)-6-fluorochromane, (1S,2R)-α-(N-Benzylaminomethyl)-6-fluoro-chroman-2-methanol |
| InChI Formula | InChI=1S/C18H20FNO2/c19-15-7-9-17-14(10-15)6-8-18(22-17)16(21)12-20-11-13-4-2-1-3-5-13/h1-5,7,9-10,16,18,20-21H,6,8,11-12H2/t16-,18+/m0/s1 |
| SMILES | O[C@@H](CNCc1ccccc1)[C@H]2CCc3cc(F)ccc3O2 |
| IUPAC Name | (1S)-2-(benzylamino)-1-[(2R)-6-fluoro-3,4-dihydro-2H-chromen-2-yl]ethanol |
| Show More |