Learn More
(1R,4S)-N-Desmethyl Sertraline Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | (1R,4S)-N-Desmethyl Sertraline Hydrochloride |
| CAS | 675126-08-6 |
| Molecular Formula | C16 H15 Cl2 N . Cl H |
| Quantity | 10 mg |
| Synonym | 1-Naphthalenamine, 4-(3,4-dichlorophenyl)-1,2,3,4-tetrahydro-, hydrochloride (1:1), (1R,4S)-, 1-Naphthalenamine, 4-(3,4-dichlorophenyl)-1,2,3,4-tetrahydro-, hydrochloride, (1R,4S)- (9CI), (1R,4S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-1-naphthalenamine hydrochloride, (1R,4S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydronaphthalen-1-amine Hydrochloride, Dasotraline hydrochloride, SEP 225289 hydrochloride, (1R,4S)-Norsertraline hydrochloride, (1R,4S)-N-Desmethyl sertraline hydrochloride, (1R,4S)-N-Desmethylsertraline hydrochloride |
| InChI Formula | InChI=1S/C16H15Cl2N.ClH/c17-14-7-5-10(9-15(14)18)11-6-8-16(19)13-4-2-1-3-12(11)13;/h1-5,7,9,11,16H,6,8,19H2;1H/t11-,16+;/m0./s1 |
| SMILES | Cl.N[C@@H]1CC[C@@H](c2ccc(Cl)c(Cl)c2)c3ccccc13 |
| IUPAC Name | (1R,4S)-4-(3,4-dichlorophenyl)-1,2,3,4-tetrahydronaphthalen-1-amine;hydrochloride |
| Molecular Weight (g/mol) | 328.66 |
| Formula Weight | 327.0348 |