Learn More
(1r,4r)-4-((2-(Butylamino)-5-(5-(morpholinomethyl)pyridin-2-yl)pyrimidin-4-yl)amino)cyclohexanol, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | (1r,4r)-4-((2-(Butylamino)-5-(5-(morpholinomethyl)pyridin-2-yl)pyrimidin-4-yl)amino)cyclohexanol |
| CAS | 1493694-70-4 |
| Molecular Formula | C24H36N6O2 |
| Quantity | 250 mg |
| Synonym | trans-4-[[2-(Butylamino)-5-[5-(4-morpholinylmethyl)-2-pyridinyl]-4-pyrimidinyl]amino]cyclohexanol, UNC2250 |
| InChI Formula | InChI=1S/C24H36N6O2/c1-2-3-10-25-24-27-16-21(23(29-24)28-19-5-7-20(31)8-6-19)22-9-4-18(15-26-22)17-30-11-13-32-14-12-30/h4,9,15-16,19-20,31H,2-3,5-8,10-14,17H2,1H3,(H2,25,27,28,29) |
| SMILES | CCCCNc1ncc(c(N[C@@H]2CC[C@@H](O)CC2)n1)c3ccc(CN4CCOCC4)cn3 |
| IUPAC Name | 4-[[2-(butylamino)-5-[5-(morpholin-4-ylmethyl)pyridin-2-yl]pyrimidin-4-yl]amino]cyclohexan-1-ol |
| Molecular Weight (g/mol) | 440.58 |