Learn More
(1R,2S,5R)-(-)-Menthyl (S)-p-Toluenesulfinate, TRC
Product Code. 30468262
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30468262 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30468262 Supplier Toronto Research Chemicals Supplier No. TRCM2204002.5G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (1R,2S,5R)-(-)-Menthyl (S)-p-Toluenesulfinate
CAS 1517-82-4
Molecular Formula C17H26O2S
Quantity 2.5 g
Synonym [1R-[1α(S*),2β,5α]]-4-Methylbenzenesulfinic Acid 5-Methyl-2-(1-methylethyl)cyclohexyl Ester, (1R,3R,4S)-Menthol (-)-(S)-p-Toluenesulfinate, (S)-p-Toluenesulfinic Acid (-)-(1R,3R,4S)-p-Menth-3-yl Ester, (-)-(1R)-Menthyl (S)-toluene-4-sulfinate
InChI Formula InChI=1S/C17H26O2S/c1-12(2)16-10-7-14(4)11-17(16)19-20(18)15-8-5-13(3)6-9-15/h5-6,8-9,12,14,16-17H,7,10-11H2,1-4H3/t14-,16+,17-,20+/m1/s1
SMILES C[C@H]1C[C@@H](O[S@@](C2=CC=C(C)C=C2)=O)[C@H](C(C)C)CC1
IUPAC Name (1R,2S,5R)-2-isopropyl-5-methylcyclohexyl (S)-4-methylbenzenesulfinate
Molecular Weight (g/mol) 294.45
Formula Weight 294.17