Learn More
SPEX Certiprep 1ML Bisabolene, 1,000 µg/mL Organic Standard
Brand:  SPEX Certiprep S-502
1ML Bisabolene, 1,000 µg/mL Organic Standard
Product Code. 16349627
  • ONLINE EXCLUSIVE PRICE
    947.00 NOK
    884.48 NOK / 1mL
    Save 62.52 NOK
    6% Off
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Bisabolene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : methanol