Learn More
SPEX Certiprep 1ML 1,2,3-Trichloropropane, 1,000 µg/mL Organic Standard
Brand:  SPEX Certiprep S-3665-MTBE
1ML 1,2,3-Trichloropropane, 1,000 µg/mL Organic Standard
Product Code. 16339457
  • ONLINE EXCLUSIVE PRICE
    871.50 NOK
    788.73 NOK / 1mL
    Save 82.77 NOK
    9% Off
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • 1,2,3-Trichloropropane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Methyl-tert-butyl ether, 1,2,3-trichloropropane