missing translation for 'onlineSavingsMsg'
Learn More

Vc-MMAD, MedChemExpress

Product Code. 16441549
Encompass_Preferred
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 16441549

Brand: Medchem Express HY15742

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vc-MMAD consists the ADCs linker (Val-Cit) and potent tubulin inhibitor (MMAD). Vc-MMAD is a drug-linker conjugate for ADC.

Vc-MMAD consists the ADCs linker (Val-Cit) and potent tubulin inhibitor (MMAD). Vc-MMAD is a drug-linker conjugate for ADC.

Specifications

Chemical Name or Material Vc-MMAD
CAS 1401963-17-4
Color White
Purity Grade Notes Research
Molecular Formula C70H104N12O14S
Quantity 1 mg
SMILES O=C1N(CCCCCC(N[C@@H](C(C)C)C(N[C@@H](CCCNC(N)=O)C(NC2=CC=C(COC(N([C@@H](C(C)C)C(N[C@@H](C(C)C)C(N([C@H]([C@@H](CC(N3[C@@H](CCC3)[C@H](OC)[C@H](C(N[C@H](C4=NC=CS4)CC5=CC=CC=C5)=O)C)=O)OC)[C@@H](C)CC)C)=O)=O)C)=O)C=C2)=O)=O)=O)C(C=C1)=O
Molecular Weight (g/mol) 1369.71
Formula Weight 1369.71
Percent Purity 98.82%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.