missing translation for 'onlineSavingsMsg'
Learn More

PSI-7409, MedChemExpress

Product Code. 16451549
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Product Code. Quantity unitSize
16451549 1 mg 1mg
16469337 5 mg 5mg
2 options
This item is not returnable. View return policy

Product Code. 16451549

Brand: Medchem Express HY15745

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress PSI-7409 is the active 5'-triphosphate metabolite of Sofosbuvir (PSI-7977). Sofosbuvir (PSI-7977) is a selective and highly active nucleotide analog inhibitor of HCV.

PSI-7409 is the active 5'-triphosphate metabolite of Sofosbuvir (PSI-7977). Sofosbuvir (PSI-7977) is a selective and highly active nucleotide analog inhibitor of HCV.

Specifications

Chemical Name or Material PSI-7409
Physical Form Solid
CAS 1015073-42-3
Color White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C10H16FN2O14P3
For Use With (Application) Cancer-programmed cell death
Solubility Information H2O : 50 mg/mL (99.97 mM; Need ultrasonic) ∣DMF : < 1 mg/mL (insoluble) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES O[C@@H]([C@@](C)(F)[C@H](N1C(NC(C=C1)=O)=O)O2)[C@H]2COP(O)(OP(OP(O)(O)=O)(O)=O)=O
Molecular Weight (g/mol) 500.16
Formula Weight 500.16
Percent Purity 98.03%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.