missing translation for 'onlineSavingsMsg'
Learn More

Niraparib metabolite M1, MedChemExpress

Product Code. 16422459
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16422459 1 mg 1mg
16411238 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16422459 Supplier Medchem Express Supplier No. HYG0023

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Niraparib metabolite M1 is a metabolite of niraparib, and the latter one acts as a novel poly(ADP-Ribose) polymerase (PARP) inhibitor.

Niraparib metabolite M1 is a metabolite of niraparib, and the latter one acts as a novel poly(ADP-Ribose) polymerase (PARP) inhibitor.

Specifications

Chemical Name or Material Niraparib metabolite M1
CAS 1476777-06-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H19N3O2
Quantity 1 mg
Synonym Niraparib carboxylic acid metabolite M1 M1 metabolite of niraparib
Solubility Information DMSO : ≥ 100 mg/mL (311.17 mM)
SMILES O=C(C1=CC=CC2=CN(C3=CC=C([C@H]4CNCCC4)C=C3)N=C12)O
Molecular Weight (g/mol) 321.37
Formula Weight 321.37
Percent Purity 99.7%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.