missing translation for 'onlineSavingsMsg'
Learn More

Nifursemizone, MedChemExpress

Product Code. 16440839
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16440839 1 mg 1mg
16496207 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16440839 Supplier Medchem Express Supplier No. HY101660
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nifursemizone is an antiprotozoal drug.

Nifursemizone is an antiprotozoal drug.

Specifications

Chemical Name or Material Nifursemizone
Physical Form Solid
CAS 5579-89-5
Color Yellow
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C8H10N4O4
For Use With (Application) COVID-19-immunoregulation
Synonym Etafurazone NF161
Solubility Information DMSO : 20.83 mg/mL (92.09 mM; ultrasonic and warming and heat to 60°C)
SMILES O=[N+](C1=CC=C(/C=N/N(C(N)=O)CC)O1)[O-]
Molecular Weight (g/mol) 226.19
Formula Weight 226.19
Percent Purity 99.25%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.