missing translation for 'onlineSavingsMsg'
Learn More

NG25, MedChemExpress

Product Code. 16451519
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Product Code. Quantity unitSize
16451519 1 mg 1mg
16489097 5 mg 5mg
16456518 10 mg 10mg
3 options
This item is not returnable. View return policy

Product Code. 16451519

Brand: Medchem Express HY15434

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NG25 is a potent dual TAK1 and MAP4K2 inhibitor, with IC50s of 149 nM and 21.7 nM, respectively.

NG25 is a potent dual TAK1 and MAP4K2 inhibitor, with IC50s of 149 nM and 21.7 nM, respectively.

Specifications

Chemical Name or Material NG25
CAS 1315355-93-1
Color White
Purity Grade Notes Research
Molecular Formula C29H30F3N5O2
Quantity 1 mg
Solubility Information DMSO : 50 mg/mL (93.01 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(NC1=CC(C(F)(F)F)=C(CN(CC2)CCN2CC)C=C1)C3=CC(OC4=C5C(NC=C5)=NC=C4)=C(C)C=C3
Molecular Weight (g/mol) 537.58
Formula Weight 537.58
Percent Purity 99.09%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.