missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-368,899 hydrochloride, MedChemExpress
Description
L-368,899 hydrochloride is a potent, selective, orally bioavailable, non-peptide oxytocin receptor antagonist, with IC50s of 8.9 nM and 26 nM for rat uterus and human uterus oxytocin receptor, respectively. L-368,899 hydrochloride used as a tocolytic agent.
Specifications
Specifications
| Chemical Name or Material | L-368,899 hydrochloride |
| CAS | 160312-62-9 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C26H43ClN4O5S2 |
| Quantity | 1 mg |
| Solubility Information | DMSO : 130 mg/mL (219.88 mM; Need ultrasonic) ∣H2O : 50 mg/mL (84.57 mM; Need ultrasonic) |
| SMILES | O=C(NC1[C@@](C2(C)C)(CS(=O)(N3CCN(C4=CC=CC=C4C)CC3)=O)CC[C@]2([H])C1)[C@H](N)CCS(=O)(C)=O.[H]Cl |
| Molecular Weight (g/mol) | 591.23 |
| Formula Weight | 591.23 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?