Learn More
MedChemExpress Dermaseptin
Specifications
Specifications
| Purity | 98.24% |
| CAS | 136212-91-4 |
| Content And Storage | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| For Use With (Application) | COVID-19-immunoregulation |
| Molecular Weight (g/mol) | 3455.1 g/mol |
| Product Type | Natural Products |
| Quantity | 1 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 25 mg/mL (7.24 mM∣Need ultrasonic) |