missing translation for 'onlineSavingsMsg'
Learn More

Chem-Lab 1LT Ammonium heptamolybdate-Sulfuric Acidreagent (50 g (NH4)6Mo7O24.4H2O + 140 ml H2SO4

Brand:  Chem-Lab CL02.0105.1000


1LT Ammonium heptamolybdate-Sulfuric Acidreagent (50 g (NH4)6Mo7O24.4H2O + 140 ml H2SO4

Product Code. 11944024

  • 1430.00 NOK / 1 L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ammonium heptamolybdate-Sulphuric Acid reagent

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contain: Ammonium heptamolybdate, Sulphuric Acid

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.