missing translation for 'onlineSavingsMsg'
Learn More

1H-Indole-3-carboxylic Acid, Mikromol™

Product Code. p-200131170
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049865 25 mg 25mg
30052099 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30049865 Supplier Mikromol Supplier No. MM0763.020025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1H-Indole-3-carboxylic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1H-Indole-3-carboxylic Acid
CAS 771-50-6
Analyte or Component Names 1H-Indole-3-carboxylic Acid
Shipping Condition Room Temperature
Molecular Formula C9 H7 N O2
Certifications/Compliance ISO 9001
Synonym Tropisetron Hydrochloride Imp. B (EP), Tropisetron Imp. B (EP), 1H-Indole-3-carboxylic Acid, Tropisetron Hydrochloride Impurity B, Tropisetron Impurity B
InChI Formula InChI=1S/C9H7NO2/c11-9(12)7-5-10-8-4-2-1-3-6(7)8/h1-5,10H,(H,11,12)
SMILES OC(=O)c1c[nH]c2ccccc12
IUPAC Name 1H-indole-3-carboxylic acid
Molecular Weight (g/mol) 161.16
Quantity 25 mg
Formula Weight 161.0477 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.