missing translation for 'onlineSavingsMsg'
Learn More

1H-Indole-2,3-dione, TRC

Product Code. 17931325
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17931325 100 g 100g
17934075 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17931325 Supplier Toronto Research Chemicals Supplier No. TRCI627060100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-56-5
InChI Formula InChI=1S/C8H5NO2/c10-7-5-3-1-2-4-6(5)9-8(7)11/h1-4H,(H,9,10,11)
IUPAC Name 1H-indole-2,3-dione
Molecular Formula C8 H5 N O2
Molecular Weight (g/mol) 147.13
Chemical Name or Material Isatin

Specifications

Quantity 100 g
Synonym Indole-2,3-dione, 2,3-Dihydro-1H-indole-2,3-dione, 2,3-Dihydroindole-2,3-dione, 2,3-Diketoindoline, 2,3-Dioxo-2,3-dihydroindole, 2,3-Dioxoindoline, 2,3-Indolindione, 2,3-Indolinedione, Isatic Acid Lactam, Isatin, Isatine, Isatinic Acid Anhydride, NSC 9262, Pseudoisatin, o-Aminobenzoylformic Anhydride
SMILES O=C1Nc2ccccc2C1=O
Formula Weight 147.032
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.