missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-Dolaphenine hydrochloride, MedChemExpress
Description
(S)-Dolaphenine hydrochloride is a component of Dolastatin 10 (HY-15580). Dolastatin 10, an antineoplastic agent, inhibits tubulin polymerization.
Specifications
Specifications
| Chemical Name or Material | (S)-Dolaphenine hydrochloride |
| Physical Form | Solid |
| CAS | 135383-60-7 |
| Color | Light Yellow |
| Quantity | 1 g |
| Purity Grade Notes | Research |
| Molecular Formula | C11H13ClN2S |
| Solubility Information | DMSO : ≥ 50 mg/mL (207.68 mM) ∣H2O : ≥ 50 mg/mL (207.68 mM) |
| SMILES | N[C@H](C1=NC=CS1)CC2=CC=CC=C2.Cl |
| Molecular Weight (g/mol) | 240.75 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?