Learn More
Thermo Scientific™ 18 m mmagnetic Screw Cap Autosampler Vial
Product Code. 15866251 Shop All Thermo Scientific Products
Click to view available options
Color:
Silver
Unit Size:
Pack of 100
Product Code. 15866251
Brand: Thermo Scientific™ 18MSCSTME
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Available in 10 mL and 20 mL usable volumes this screw thread vial requires no tools for closure. Magnetic caps are ideal for magnetic pick up in autosamplers.
TRUSTED_SUSTAINABILITY
Specifications
Color (Seal) White
Color Silver
Material Steel
Septum Blue Silicone/Clear PTFE
For Use With (Application) Solid Phase Micro Extraction (SPME)
Thread Style HS
Product Line Chromacol
Quantity 100
Thickness (Metric) 0.2 mm
Type Screw Cap (Magnetic)
Closure Size 18 mm