Learn More
Bochem™ 18/10 Stainless Steel Standard Spoon Spatula
Product Code. 10045070
Change view
Click to view available options
Length (Metric):
230 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10045070 230 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10045070 Supplier Bochem™ Supplier No. 3215
Estimated Shipment: 04-01-2027
Manufactured from 18/10 Stainless Steel. BOCHEM™ 18/10 Stainless Steel Standard Spoon Spatula is a standard spatula.
  •  Stainless steel
Specifications
Quantity 1 Ea.
Autoclavable Autoclavable
Product Type Spoon Spatula
For Use With (Application) Designed for a wide range of uses
Diameter (Metric) Outer 4mm
Material Stainless Steel (18/10)
Length (Metric) 230 mm