Learn More
Bochem™ 18/10 Stainless Steel Spoon Spatulas
Product Code. 11875071
Change view
Click to view available options
Length (Metric):
150 mm
180 mm
210 mm
250 mm
300 mm
Length (Metric) Blade:
40 mm
50 mm
70 mm
Unit Size:
Each
Pack of 10
Product Code. Length (Metric) Length (Metric) Blade unitSize
11875071 210 mm 50 mm Pack of 10
11825151 150 mm 40 mm Pack of 10
10767391 180 mm 50 mm Each
10579851 250 mm 70 mm Each
10303571 300 mm 70 mm Each
5 options
Product Code. 11875071
Brand: Bochem™ 3473
Only null left
Add to basket
Add to basket
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Spoon Spatulas serve a wide variety of functions in the lab.
  • Manufactured from 18/10 stainless steel
Specifications
Quantity 10 Pk.
Product Type Spoon Spatula
Length (Metric) Blade 50 mm
Width (Metric) 12 mm
Width (Metric) Blade 10 mm
Material Stainless Steel (18/10)
Length (Metric) 210 mm