Learn More
Bochem™ 18/10 Stainless Steel Spatulas, With knob
Product Code. 10706821
Click to view available options
Length (Metric):
150 mm
210 mm
300 mm
Diameter (Metric) Outer:
4mm
4.5mm
5mm
Unit Size:
Each
Product Code. 10706821
Brand: Bochem™ 3270
Only null left
Add to basket
Add to basket
Manufactured from 18/10 stainless steel
  • Made from 18/10 stainless steel
Specifications
Quantity 1 Ea.
Shape With Knob
Product Type Spatula
Length (Metric) Blade 60 mm
Width (Metric) 9 mm
Width (Metric) Blade 9 mm
Blade Material Stainless Steel (18/10)
Diameter (Metric) Outer 4mm
Dimensions (L x W) 150 x 9 mm
Material Stainless Steel (18/10)
Length (Metric) 150 mm