Learn More
Bochem™ 18/10 Stainless Steel Spatulas, Spoon/round/bent
Product Code. 10494291
Change view
Click to view available options
Length (Metric):
180 mm
185 mm
210 mm
Diameter (Metric) Outer:
2mm
3mm
4mm
Unit Size:
Each
Product Code. Length (Metric) Diameter (Metric) Outer unitSize
10494291 185 mm 2mm Each
10335671 210 mm 4mm Each
10579422 180 mm 3mm Each
10663541 210 mm 2mm Each
4 options
Product Code. 10494291
missing translation for 'mfr': Bochem™ 3201
Only null left
Add to basket
Add to basket
Manufactured from 18/10 stainless steel
  • Made from 18/10 stainless steel
Specifications
Quantity 1 Ea.
Autoclavable Autoclavable
Shape Spoon/Round/Bent
Product Type Spatula
For Use With (Application) Used for a variety of applications
Blade Material Stainless Steel (18/10)
Diameter (Metric) Outer 2mm
Material Stainless Steel (18/10)
Length (Metric) 185 mm