Learn More
Bochem™ 18/10 Stainless Steel Section Lifters, with Wooden Handle
Product Code. 11815181
Change view
Click to view available options
Dimensions (L x W):
150 x 10 mm
150 x 15 mm
Unit Size:
Pack of 10
Product Code. Dimensions (L x W) unitSize
11815181 150 x 10 mm Pack of 10
11885051 150 x 15 mm Pack of 10
2 options
Product Code. 11815181
Brand: Bochem™ 3535
Only null left
Add to basket
Add to basket
Manufactured from 18/10 stainless steel
Specifications
Quantity 1 Ea.
Product Type Section Lifter
Length (Metric) Blade 50 mm
Width (Metric) 10 mm
Blade Material Stainless Steel (18/10)
Diameter (Metric) Outer 0.4mm
Dimensions (L x W) 150 x 10 mm
Handle Material Wood
Material Stainless Steel (18/10)
Length (Metric) 150 mm