Learn More
Bochem™ 18/10 Stainless Steel Scoop Spatula
Description
- 1 side of spatula is tapered
- Easy grip
- Stainless steel
- Analyse type
Specifications
Specifications
| Quantity | 1 Ea. |
| Autoclavable | Autoclavable |
| Shape | Spoon |
| Product Type | Spoon Spatula |
| For Use With (Application) | For analysis |
| Dimensions (L x W) | F x E 35 x 23 |
| Material | Stainless Steel (18/10) |
| Handle Type | Tapered |
| Length (Metric) | 200 mm |