Learn More
Bochem™ 18/10 Stainless Steel Poly Spoons
Product Code. 10370481
Change view
Click to view available options
Length (Metric):
150 mm
180 mm
190 mm
210 mm
250 mm
300 mm
400 mm
500 mm
Length (Metric) Blade:
26 mm
30 mm
35 mm
Unit Size:
Each
Product Code. Length (Metric) Length (Metric) Blade unitSize
10370481 210 mm 35 mm Each
10219130 150 mm 30 mm Each
10370301 180 mm 30 mm Each
10211351 190 mm 26 mm Each
10248800 250 mm 30 mm Each
10791601 300 mm 35 mm Each
10506562 300 mm 30 mm Each
10675601 400 mm 35 mm Each
10050771 500 mm 35 mm Each
9 options
Product Code. 10370481
Brand: Bochem™ 3410
Only null left
Add to basket
Add to basket
Manufactured from 18/10 stainless steel
  • Spoon comes in three different types
  • Autoclavable
  • Made of 18/10 stainless steel
  • Multi-purpose spoon for a wide range of uses
Specifications
Quantity 1 Ea.
Autoclavable Autoclavable
Product Type Poly Spoon
For Use With (Application) Multi-purpose spoon for a wide range of uses
Length (Metric) Blade 35 mm
Width (Metric) 20 mm
Blade Material Stainless Steel (18/10)
Diameter (Metric) Outer 8mm
Dimensions (L x W) 210 x 20 mm
Material Stainless Steel (18/10)
Length (Metric) 210 mm