Learn More
Bochem™ 18/10 Stainless Steel Poly-Spoon
Product Code. 11845071
Change view
Click to view available options
Length (Metric):
210 mm
Unit Size:
Pack of 10
Product Code. Length (Metric) unitSize
11845071 210 mm Pack of 10
1 options
Product Code. 11845071
Brand: Bochem™ 3402
Only null left
Add to basket
Add to basket
Made from 18/10 Stainless Steel.
  • Type 1
  • Stainless Steel
Specifications
Shape Bent
Product Type Poly Spoon
Width (Metric) 13 mm
Width (Metric) Blade 13 mm
Material Stainless Steel (18/10)
Length (Metric) 210 mm