Learn More
Bochem™ 18/10 Stainless Steel Pharma Scoop
1395.00 NOK - 1940.00 NOK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10000861
|
Bochem™
8813 |
2 L |
1940.00 NOK
Each |
Estimated Shipment: 25-04-2024
Log in to see stock. |
|||||
10458432
|
Bochem™
8810 |
250 mL |
1395.00 NOK
Each |
Estimated Shipment: 02-05-2024
Log in to see stock. |
|||||
10000791
|
Bochem™
8811 |
500 mL |
1500.00 NOK
Each |
Estimated Shipment: 29-04-2024
Log in to see stock. |
|||||
10468432
|
Bochem™
8812 |
1000 mL |
1675.00 NOK
Each |
Estimated Shipment: 29-04-2024
Log in to see stock. |
|||||
Specifications
Pharma Scoop | |
Stainless Steel (18/10) |