Learn More
Bochem™ 18/10 Stainless Steel One-Sided Chemical Spoons
Description
- One-sided chemical spoon
- 18/10 stainless steel
Specifications
Specifications
| Quantity | 1 Ea. |
| Product Type | Chemical Scoop |
| Diameter (Metric) Outer | 5mm |
| Handle Material | Stainless Steel (18/10) |
| Material | Stainless Steel (18/10) |
| Length (Metric) | 180 mm |