Learn More
Bochem™ 18/10 Stainless Steel Micro Spoon Spatulas, Spoon Shape
Product Code. 10721202
Change view
Click to view available options
Length (Metric):
130 mm
180 mm
185 mm
210 mm
Product Type:
Micro Spoon Spatula
Spoon Spatula
Diameter (Metric) Outer:
2mm
2.5mm
3mm
4mm
Unit Size:
Each
Pack of 10
Product Code. Length (Metric) Product Type Diameter (Metric) Outer unitSize
10721202 180 mm Spoon Spatula 3mm Each
10420441 210 mm Micro Spoon Spatula 2mm Each
10770812 130 mm Micro Spoon Spatula 2mm Each
10094950 180 mm Micro Spoon Spatula 2.5mm Each
11835071 185 mm Micro Spoon Spatula 2mm Pack of 10
10557122 210 mm Spoon Spatula 4mm Each
6 options
Product Code. 10721202
Brand: Bochem™ 3213
Only null left
Add to basket
Add to basket
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Micro Spoon Spatulas comes in two types.
  • Spoon spatula
  • 18/10 stainless steel
Specifications
Quantity 1 Ea.
Shape Spoon
Product Type Spoon Spatula
Diameter (Metric) Outer 3mm
Material Stainless Steel (18/10)
Length (Metric) 180 mm