Learn More
Bochem™ 18/10 Stainless Steel Micro-Spatula
Product Code. 10765073
Change view
Click to view available options
Length (Metric):
160 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
10765073 160 mm Each
1 options
Product Code. 10765073
Brand: Bochem™ 3231
Only null left
Add to basket
Add to basket
Manufactured from 18/10 stainless steel. BOCHEM™ 18/10 Stainless Steel Micro-Spatula has a PVC handle.
  • 18/10 stainless steel
  • PVC handle
Specifications
Quantity 1 Ea.
Shape Rounded Flat
Product Type Micro Spatula
Length (Metric) Blade 40 mm
Width (Metric) 4 mm
Width (Metric) Blade 4 mm
Handle Material PVC
Material Stainless Steel (18/10)
Length (Metric) 160 mm