Learn More
Bochem™ 18/10 Stainless Steel Micro Scoop
Product Code. 10752352
Change view
Click to view available options
Dimensions (L x W):
210 x 4 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
10752352 210 x 4 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10752352 Supplier Bochem™ Supplier No. 3306
Manufactured from 18/10 Stainless Steel. BOCHEM™ 18/10 Stainless Steel Micro Scoop had a wide scoop.
  • Micro scoop
  • Half-rounded
  • Wide
Specifications
Product Type Micro Scoop
Width (Metric) 4 mm
Dimensions (L x W) 210 x 4 mm
Material Stainless Steel (18/10)
Length (Metric) 210 mm