Learn More
Bochem™ 18/10 Stainless Steel Measure Spoon Set
Description
- 18/10 stainless steel
- Four sizes:
- 1.0ml
- 2.5ml
- 5ml
- 15ml
Specifications
Specifications
| Quantity | 1 Ea. |
| Capacity (Metric) | 1 mL, 2.5 mL, 5 mL, 15 mL |
| Product Type | Measure Spoon Set |
| Material | Stainless Steel (18/10) |