Learn More
Bochem™ 18/10 Stainless Steel Laboratory Spoons
Description
- Durable 18/10 stainless steel construction.
- Polished surface for easy cleaning.
- Oval bowl for easy material pickup and delivery.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Product Type | Laboratory Spoon |
| For Use With (Application) | Multi-purpose spoon for a wide range of uses |
| Diameter (Metric) Outer | 10mm |
| Material | Stainless Steel (18/10) |
| Length (Metric) | 150 mm |