Learn More
Bochem™ 18/10 Stainless Steel Laboratory Spoon
Product Code. 10673072
Change view
Click to view available options
Length (Metric):
150 mm
170 mm
200 mm
Dimensions (L x W):
20 x 10 mm
30 x 15 mm
30 x 26 mm
40 x 10 mm
Unit Size:
Each
Product Code. Length (Metric) Dimensions (L x W) unitSize
10673072 150 mm 30 x 15 mm Each
10176360 150 mm 20 x 10 mm Each
10298790 200 mm 30 x 26 mm Each
10454871 170 mm 40 x 10 mm Each
4 options
Product Code. 10673072
Brand: Bochem™ 3380
Only null left
Add to basket
Edge
Add to basket
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Laboratory Spoon serve a wide variety of functions in the lab.
  • Lab spoon
  • Manufactured from 18/10 stainless steel
Specifications
Quantity 1 Ea.
Product Type Laboratory Spoon
Dimensions (L x W) 30 x 15 mm
Material Stainless Steel (18/10)
Length (Metric) 150 mm