Learn More
Bochem™ 18/10 Stainless Steel Lab Jacks
Product Code. 10408632
Change view
Click to view available options
Width (Metric):
100 mm
130 mm
200 mm
240 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) unitSize
10408632 100 mm Each
10088521 200 mm Each
10303712 130 mm Each
10258640 240 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10408632 Supplier Bochem™ Supplier No. 11115
Estimated Shipment: 14-09-2026
Specifications
Material Stainless Steel (18/10)
Dimensions (L x W) 100 x 100 mm
Length (Metric) Base 100 mm
Width (Metric) 100 mm
Includes Wheels
Product Type Lab Jack
Quantity 1 Ea.