Learn More
Bochem™ 18/10 Stainless Steel High Shape Laboratory Bowl
Product Code. 10103504
Click to view available options
Capacity (Metric):
1.5 L
2 L
3 L
6 L
8 L
10 L
Diameter (Metric):
150 mm
180 mm
195 mm
220 mm
285 mm
310 mm
Height (Metric):
80 mm
95 mm
115 mm
125 mm
145 mm
165 mm
Unit Size:
Each
Product Code. 10103504
Brand: Bochem™ 8601
Manufactured from 18/10 stainless steel
  • High shape
  • With rim
  • 18/10 stainless steel
Specifications
Bottom Shape Flat
Capacity (Metric) 10 L
Material Stainless Steel
Shape Round
Type Laboratory Bowl
Height (Metric) 80 mm
Diameter (Metric) 150 mm