Learn More
Bochem™ 18/10 Stainless Steel Guide Roll
Product Code. 10374081
Change view
Click to view available options
Load Bearing Capacity (Metric):
70 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Load Bearing Capacity (Metric) Unit Size
10374081 70 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10374081 Supplier Bochem™ Supplier No. 100
Estimated Shipment: 01-03-2027
Specifications
Diameter (Metric) 100 mm
Height (Metric) 121 mm
Load Bearing Capacity (Metric) 70 kg
Material 18/10 Stainless Steel
Width (English) 3/4 in.
Width (Metric) 26.9 mm