Learn More
Bochem™ 18/10 Stainless Steel Evaporating Dish with Rim
Product Code. 10098310
Click to view available options
Height (Metric):
45 mm
60 mm
95 mm
100 mm
Length (Metric):
310 mm
340 mm
375 mm
445 mm
500 mm
Unit Size:
Each
Product Code. 10098310
Brand: Bochem™ 8456
Only null left
Add to basket
Add to basket
Evaporate materials in the laboratory wit the Bochem™ 18/10 Stainless Steel Evaporating Dish with Rim.
  • 18/10 stainless steel
  • Substantial rim size
  • Rectangular shape
Specifications
Material Stainless Steel (18/10)
Type Evaporating Dish
Length (Metric) 310 mm
Height (Metric) 45 mm