Learn More
Bochem™ 18/10 Stainless Steel Evaporating Dish
Specifications
Specifications
| Bottom Shape | Flat |
| Capacity (Metric) | 50 mL |
| Material | Stainless Steel |
| Shape | Round |
| Type | Evaporating Dish |
| Height (Metric) | 20 mm |
| Diameter (Metric) | 60 mm |