Learn More
Bochem™ 18/10 Stainless Steel Double Spatulas, Straight
Description
- Double spatula
- Manufactured from 18/10 stainless steel
Specifications
Specifications
| Quantity | 1 Ea. |
| Shape | Straight |
| Coating | PTFE |
| Product Type | Double Ended Spatula |
| Length (Metric) Blade | 60 mm |
| Width (Metric) | 11 mm |
| Width (Metric) Blade | 11 mm |
| Material | Stainless Steel (18/10) |
| Length (Metric) | 210 mm |